Assay Details
Target Gene Details
Entrez Gene ID: | 26037 |
Gene Name: | signal induced proliferation associated 1 like 1 |
Gene Aliases: |
E6TP1 |
Location: |
Chr.14:71320418-71741229 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv832827 | Chr.14:71209461 - 71366802 on Build GRCh38 | Gain | LOC105370706 SIPA1L1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |