Assay Details
Target Gene Details
Entrez Gene ID: | 4738 |
Gene Name: | neural precursor cell expressed, developmentally down-regulated 8 |
Gene Aliases: |
NEDD-8 |
Location: |
Chr.14:24216851-24232370 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NEDD8 | NM_006156.2 | NP_006147.1 | ||
AK125214.1 | ||||
BC104200.1 | AAI04201.1 | |||
BC104201.1 | AAI04202.1 | |||
BC104664.1 | AAI04665.1 | |||
BE548741.1 | ||||
CF130271.1 | ||||
CR407662.1 | CAG28590.1 | |||
D23662.1 | BAA04889.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic non-DGV Variation |