Assay Details
Target Gene Details
Entrez Gene ID: | 143 |
Gene Name: | poly(ADP-ribose) polymerase family member 4 |
Gene Aliases: |
ADPRTL1, ARTD4, PARP-4, PARPL, PH5P, VAULT3, VPARP, VWA5C, p193 |
Location: |
Chr.13:24420930-24512993 on Build GRCh38 |
Assay Gene Location: | Within Intron 25 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PARP4 | NM_006437.3 | NP_006428.2 | ||
XM_011534931.1 | XP_011533233.1 | |||
XM_011534932.2 | XP_011533234.1 | |||
AF057160.1 | AAC62491.1 | |||
AF158255.1 | AAD47250.1 | |||
D79999.2 | ||||
T10289.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv832567 | Chr.13:24302981 - 24520733 on Build GRCh38 | Loss | PARP4 NUS1P3 C1QTNF9 LINC00566 SPATA13 |
esv3892310 | Chr.13:24212887 - 24577193 on Build GRCh38 | Gain | LOC101927375 PARP4 NUS1P3 C1QTNF9 SPATA13-AS1 LINC00566 LOC105370295 SPATA13 |
nsv1036613 | Chr.13:24347567 - 24461469 on Build GRCh38 | Gain | PARP4 |
nsv1048909 | Chr.13:24438079 - 24465528 on Build GRCh38 | Gain | PARP4 |
dgv1616n100 | Chr.13:24367378 - 24455157 on Build GRCh38 | Gain | PARP4 |
dgv1617n100 | Chr.13:24445874 - 24467049 on Build GRCh38 | Loss | PARP4 |
esv3582215 | Chr.13:24193682 - 24455263 on Build GRCh38 | Gain | PARP4 NUS1P3 C1QTNF9 SPATA13-AS1 LINC00566 SPATA13 |
esv3631556 | Chr.13:24426828 - 24487208 on Build GRCh38 | Loss | PARP4 |
esv2761200 | Chr.13:24313097 - 24636778 on Build GRCh38 | Gain+Loss | LOC101927375 PARP4 NUS1P3 C1QTNF9 LINC00566 TPTE2P6 LOC105370295 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |