Assay Details
Target Gene Details
Entrez Gene ID: | 338811 |
Gene Name: | family with sequence similarity 19 member A2, C-C motif chemokine like |
Gene Aliases: |
TAFA-2, TAFA2 |
Location: |
Chr.12:61708248-62192839 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
FAM19A2 | NM_178539.4 | NP_848634.1 | ||
XM_006719370.2 | XP_006719433.1 | |||
XM_011538272.1 | XP_011536574.1 | |||
XM_011538274.1 | XP_011536576.1 | |||
AK123580.1 | ||||
AL834160.1 | CAD38865.1 | |||
AY325115.1 | AAP92407.1 | |||
BC028403.1 | AAH28403.1 | |||
BC040286.1 | AAH40286.1 | |||
CR749367.1 | CAH18220.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3581660 | Chr.12:61685385 - 61733005 on Build GRCh38 | Gain | FAM19A2 |
nsv1042044 | Chr.12:60862234 - 61803104 on Build GRCh38 | Gain | FAM19A2 |
nsv520898 | Chr.12:61684989 - 61721384 on Build GRCh38 | Gain | FAM19A2 |
nsv1039741 | Chr.12:61653069 - 62097271 on Build GRCh38 | Loss | FAM19A2 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |