Assay Details
Target Gene Details
Entrez Gene ID: | 55967 |
Gene Name: | NADH:ubiquinone oxidoreductase subunit A12 |
Gene Aliases: |
B17.2, DAP13 |
Location: |
Chr.12:94971328-95003713 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NDUFA12 | NM_001258338.1 | NP_001245267.1 | ||
NM_018838.4 | NP_061326.1 | |||
AF112208.1 | AAF17196.1 | |||
AF217092.1 | AAF91224.1 | |||
AK222619.1 | BAD96339.1 | |||
BC005936.1 | AAH05936.1 | |||
BG943528.1 | ||||
BT007220.1 | AAP35884.1 | |||
BU532909.1 | ||||
DW441120.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic non-DGV Variation |