Assay Details
Target Gene Details
Entrez Gene ID: | 140707 |
Gene Name: | BRI3 binding protein |
Gene Aliases: |
BNAS1, HCCR-1, HCCR-2, HCCRBP-1, KG19 |
Location: |
Chr.12:124993648-125051845 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
BRI3BP | NM_080626.5 | NP_542193.3 | ||
XM_011537940.2 | XP_011536242.1 | |||
AF283671.2 | AAL33003.2 | |||
AF284094.1 | AAL54382.1 | |||
AF441865.1 | AAN76689.1 | |||
BC034525.1 | AAH34525.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1051020 | Chr.12:125015140 - 125809096 on Build GRCh38 | Gain | TMEM132B THRIL BRI3BP AACS |
nsv1047470 | Chr.12:125009120 - 125044073 on Build GRCh38 | Loss | THRIL BRI3BP |
nsv560595 | Chr.12:124737381 - 125019207 on Build GRCh38 | Gain | MIR5188 SCARB1 TRF-GAA1-4 DHX37 BRI3BP UBC |
nsv509485 | Chr.12:124990941 - 125025754 on Build GRCh38 | Insertion | THRIL BRI3BP |