Assay Details
Target Gene Details
Entrez Gene ID: | 9380 |
Gene Name: | glyoxylate reductase/hydroxypyruvate reductase |
Gene Aliases: |
GLXR, GLYD, PH2 |
Location: |
Chr.9:37422666-37437782 on Build GRCh38 |
Assay Gene Location: | Within Intron 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
GRHPR | NM_012203.1 | NP_036335.1 | ||
XM_005251631.1 | XP_005251688.1 | |||
XM_011518073.2 | XP_011516375.1 | |||
XM_017015320.1 | XP_016870809.1 | |||
XM_017015321.1 | XP_016870810.1 | |||
XM_017015322.1 | XP_016870811.1 | |||
XM_017015323.1 | XP_016870812.1 | |||
AF113215.1 | AAG39286.1 | |||
AF113251.1 | AAD54066.1 | |||
AF134895.1 | AAF00111.1 | |||
AF146018.1 | AAD45886.1 | |||
AK024386.1 | ||||
AK025978.1 | ||||
AK026287.1 | BAB15430.1 | |||
AK026752.1 | ||||
AK315690.1 | ||||
BC000605.2 | AAH00605.1 | |||
BC088360.1 | AAH88360.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic non-DGV Variation |