Assay Details
Target Gene Details
Entrez Gene ID: | 105378159 |
Gene Name: | translation initiation factor IF-2-like |
Gene Aliases: |
- |
Location: |
Chr.6:130288003-130386905 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LOC105378159 | XM_017011599.1 | XP_016867088.1 |
Target Gene Details
Entrez Gene ID: | 154075 |
Gene Name: | sterile alpha motif domain containing 3 |
Gene Aliases: |
- |
Location: |
Chr.6:130133203-130365425 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SAMD3 | NM_001258275.2 | NP_001245204.1 | ||
AK091351.1 | BAC03644.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv604663 | Chr.6:129482340 - 130671010 on Build GRCh38 | Gain | TMEM200A TMEM244 LAMA2 SAMD3 ARHGAP18 LOC102723409 LOC105378159 L3MBTL3 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |