Assay Details
Target Gene Details
Entrez Gene ID: | 64332 |
Gene Name: | NFKB inhibitor zeta |
Gene Aliases: |
IKBZ, INAP, MAIL |
Location: |
Chr.3:101827990-101861025 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NFKBIZ | NM_001005474.2 | NP_001005474.1 | ||
AB074898.1 | ||||
AF548362.1 | AAN40698.1 | |||
AK054787.1 | ||||
BM050734.1 | ||||
DA541852.1 | ||||
DQ224339.1 | ABB02425.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv953238 | Chr.3:101825757 - 101845056 on Build GRCh38 | Duplication | NXPE3 NFKBIZ |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |