Assay Details
Target Gene Details
Entrez Gene ID: | 201562 |
Gene Name: | 3-hydroxyacyl-CoA dehydratase 2 |
Gene Aliases: |
PTPLB |
Location: |
Chr.3:123491548-123585093 on Build GRCh38 |
Assay Gene Location: | Within Intron 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
HACD2 | NM_198402.3 | NP_940684.1 | ||
XM_017005855.1 | XP_016861344.1 | |||
AK074605.1 | ||||
AK289983.1 | ||||
AL832532.1 | ||||
AY191814.1 | AAP20101.1 | |||
BC049369.1 | AAH49369.1 | |||
BC060839.1 | AAH60839.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv829707 | Chr.3:123357546 - 123540363 on Build GRCh38 | Loss | HACD2 ADCY5 |
nsv3978 | Chr.3:123456399 - 123501453 on Build GRCh38 | Deletion | HACD2 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |