Assay Details
Target Gene Details
Entrez Gene ID: | 2882 |
Gene Name: | glutathione peroxidase 7 |
Gene Aliases: |
CL683, GPX6, GPx-7, GSHPx-7, NPGPx |
Location: |
Chr.1:52602371-52609051 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
GPX7 | NM_015696.4 | NP_056511.2 | ||
AF320068.2 | AAN76501.1 | |||
AK027683.1 | BAB55294.1 | |||
AY358402.1 | AAQ88768.1 | |||
BC032788.1 | AAH32788.1 | |||
BI550170.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic non-DGV Variation |