Assay Details
Target Gene Details
Entrez Gene ID: | 10753 |
Gene Name: | calpain 9 |
Gene Aliases: |
GC36, nCL-4 |
Location: |
Chr.1:230747289-230803356 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CAPN9 | NM_001319676.1 | NP_001306605.1 | ||
NM_006615.2 | NP_006606.1 | |||
NM_016452.2 | NP_057536.1 | |||
XM_011544017.1 | XP_011542319.1 | |||
XM_011544018.1 | XP_011542320.1 | |||
XM_011544019.2 | XP_011542321.1 | |||
XM_011544020.1 | XP_011542322.1 | |||
XM_017000098.1 | XP_016855587.1 | |||
XM_017000099.1 | XP_016855588.1 | |||
AB038463.1 | BAA92137.1 | |||
AF022799.1 | AAB80762.1 | |||
AK298024.1 | ||||
AK309039.1 | ||||
AK313629.1 | ||||
BC027993.1 | AAH27993.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3589093 | Chr.1:230482037 - 231277190 on Build GRCh38 | Gain | LOC101927553 ARV1 FAM89A AGT TRIM67 MIR1182 C1orf131 C1orf198 COG2 GNPAT LOC101927604 LOC149373 TTC13 CAPN9 |
nsv549335 | Chr.1:230733804 - 230797361 on Build GRCh38 | Gain | CAPN9 |
nsv832836 | Chr.1:230693328 - 230832263 on Build GRCh38 | Loss | COG2 AGT CAPN9 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |