Assay Details
Target Gene Details
Entrez Gene ID: | 100505592 |
Gene Name: | gastric adenocarcinoma associated, positive CD44 regulator, long intergenic non-coding RNA |
Gene Aliases: |
LINC01540 |
Location: |
Chr.18:3466250-3478978 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
GAPLINC | NR_110428.1 | |||
NR_110429.1 | ||||
AJ707365.1 | ||||
BM782281.1 | ||||
BX107736.1 |