Assay Details
Target Gene Details
Entrez Gene ID: | 793 |
Gene Name: | calbindin 1 |
Gene Aliases: |
CALB, D-28K |
Location: |
Chr.8:90058608-90082881 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CALB1 | NM_004929.3 | NP_004920.1 | ||
AK054881.1 | ||||
AK295585.1 | ||||
AK312491.1 | ||||
AK315959.1 | ||||
BC006478.1 | AAH06478.1 | |||
BC020864.1 | AAH20864.1 | |||
HY128922.1 | ||||
X06661.1 | CAA29860.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2759628 | Chr.8:89988558 - 90317441 on Build GRCh38 | Gain+Loss | LINC00534 CALB1 DECR1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |