Assay Details
Target Gene Details
Entrez Gene ID: | 84456 |
Gene Name: | l(3)mbt-like 3 (Drosophila) |
Gene Aliases: |
MBT-1, MBT1 |
Location: |
Chr.6:130018583-130141449 on Build GRCh38 |
Assay Gene Location: | Within Intron 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
L3MBTL3 | NM_001007102.2 | NP_001007103.1 | ||
NM_032438.2 | NP_115814.1 | |||
XM_005267161.4 | XP_005267218.1 | |||
XM_006715576.3 | XP_006715639.1 | |||
XM_006715578.3 | XP_006715641.1 | |||
XM_011536179.2 | XP_011534481.1 | |||
XM_011536180.2 | XP_011534482.1 | |||
XM_011536181.2 | XP_011534483.1 | |||
XM_011536183.2 | XP_011534485.1 | |||
XM_011536184.2 | XP_011534486.1 | |||
XM_017011355.1 | XP_016866844.1 | |||
XM_017011356.1 | XP_016866845.1 | |||
AB058701.1 | ||||
AK094137.1 | ||||
AK290219.1 | ||||
BC060845.1 | AAH60845.1 | |||
DA927626.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv604663 | Chr.6:129482340 - 130671010 on Build GRCh38 | Gain | LAMA2 TMEM200A LOC102723409 LOC105378159 TMEM244 ARHGAP18 SAMD3 L3MBTL3 |
esv3610778 | Chr.6:129848176 - 130076578 on Build GRCh38 | Gain | TMEM244 L3MBTL3 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |