Assay Details
Target Gene Details
Entrez Gene ID: | 4338 |
Gene Name: | molybdenum cofactor synthesis 2 |
Gene Aliases: |
MCBPE, MOCO1, MOCODB, MPTS |
Location: |
Chr.5:53095679-53109772 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
MOCS2 | NM_004531.4 | NP_004522.1 | ||
NM_176806.3 | NP_789776.1 | |||
AF091871.1 | AAD14598.1 | |||
AF117815.1 | AAD13296.1 | |||
AF155659.1 | AAF67478.1 | |||
AK126211.1 | ||||
AK312887.1 | ||||
BC046097.1 | AAH46097.1 | |||
CD514049.1 | ||||
CR457172.1 | CAG33453.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv830296 | Chr.5:52913698 - 53114423 on Build GRCh38 | Gain | MOCS2 LOC257396 ITGA2 ITGA1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |