Assay Details
Target Gene Details
Entrez Gene ID: | 7015 |
Gene Name: | telomerase reverse transcriptase |
Gene Aliases: |
CMM9, DKCA2, DKCB4, EST2, PFBMFT1, TCS1, TP2, TRT, hEST2, hTRT |
Location: |
Chr.5:1253167-1295626 on Build GRCh38 |
Assay Gene Location: | Within Intron 13 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TERT | NM_001193376.1 | NP_001180305.1 | ||
NM_198253.2 | NP_937983.2 | |||
XM_017009796.1 | XP_016865285.1 | |||
AB085628.1 | BAC11010.1 | |||
AB086379.1 | BAC11014.1 | |||
AB086950.1 | BAC11015.1 | |||
AF015950.1 | AAC51672.1 | |||
AF018167.1 | AAC51724.1 | |||
AX810036.1 | CAE75638.1 | |||
AX810378.1 | CAE75644.1 | |||
JF896281.1 | ||||
JF896283.1 | ||||
JF896284.1 | ||||
JF896285.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1024507 | Chr.5:1232783 - 1269250 on Build GRCh38 | Gain |
![]() ![]() |
esv3575810 | Chr.5:1055511 - 1282578 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() |
nsv509044 | Chr.5:1147636 - 1265572 on Build GRCh38 | Insertion |
![]() ![]() ![]() ![]() ![]() |
nsv950627 | Chr.5:1008886 - 1300685 on Build GRCh38 | Deletion |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv596629 | Chr.5:457146 - 1468172 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
esv3603797 | Chr.5:1086744 - 1259707 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)