Assay Details
Target Gene Details
Entrez Gene ID: | 27306 |
Gene Name: | hematopoietic prostaglandin D synthase |
Gene Aliases: |
GSTS, GSTS1, GSTS1-1, PGD2, PGDS |
Location: |
Chr.4:94298556-94342876 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
HPGDS | NM_014485.2 | NP_055300.1 | ||
XM_005262932.2 | XP_005262989.1 | |||
AK290075.1 | ||||
BC020734.1 | AAH20734.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv594901 | Chr.4:94333579 - 94572095 on Build GRCh38 | Gain | HPGDS PDLIM5 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |