Assay Details
Target Gene Details
Entrez Gene ID: | 253017 |
Gene Name: | trans-2,3-enoyl-CoA reductase-like |
Gene Aliases: |
GPSN2L, SRD5A2L2, TERL |
Location: |
Chr.4:64276298-64409509 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TECRL | NM_001010874.4 | NP_001010874.2 | ||
XM_005265662.4 | XP_005265719.1 | |||
XM_005265663.2 | XP_005265720.1 | |||
XM_005265664.2 | XP_005265721.1 | |||
XM_005265665.3 | XP_005265722.1 | |||
XM_017007959.1 | XP_016863448.1 | |||
XM_017007960.1 | XP_016863449.1 | |||
AK290606.1 | ||||
AL833108.1 | CAI46109.1 | |||
BC072463.1 | AAH72463.1 | |||
BX647590.1 | CAI46063.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv594472 | Chr.4:64292373 - 64537688 on Build GRCh38 | Gain | TECRL |
nsv998669 | Chr.4:63850701 - 64580449 on Build GRCh38 | Loss | TECRL |
nsv1012062 | Chr.4:64277750 - 64318606 on Build GRCh38 | Loss | TECRL |
nsv594372 | Chr.4:62968251 - 65067958 on Build GRCh38 | Loss | LOC401134 TECRL |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |