Assay Details
Target Gene Details
Entrez Gene ID: | 23171 |
Gene Name: | glycerol-3-phosphate dehydrogenase 1-like |
Gene Aliases: |
GPD1-L |
Location: |
Chr.3:32106511-32168715 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
GPD1L | NM_015141.3 | NP_055956.1 | ||
XM_006713068.2 | XP_006713131.1 | |||
AK292808.1 | ||||
BC028726.2 | AAH28726.1 | |||
D42047.1 | ||||
DA318838.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1003285 | Chr.3:31386933 - 33172886 on Build GRCh38 | Gain | OSBPL10-AS1 LOC105377021 CMTM7 GLB1 STT3B GPD1L LOC105377022 CMTM8 TMPPE ZNF860 DYNC1LI1 MIR548AY TRIM71 SUSD5 CRTAP CNOT10 CCR4 OSBPL10 CMTM6 |
nsv1011573 | Chr.3:31860029 - 32251734 on Build GRCh38 | Gain | GPD1L CMTM8 OSBPL10 ZNF860 |
nsv3751 | Chr.3:32088214 - 32133605 on Build GRCh38 | Deletion | GPD1L |
nsv1003110 | Chr.3:32101304 - 32140890 on Build GRCh38 | Gain | GPD1L |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |