Assay Details
Target Gene Details
Entrez Gene ID: | 101928371 |
Gene Name: | uncharacterized LOC101928371 |
Gene Aliases: |
- |
Location: |
Chr.2:88538720-88575610 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LOC101928371 | NR_110236.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv470476 | Chr.2:88384248 - 88829483 on Build GRCh38 | Gain | ANKRD36BP2 FOXI3 EIF2AK3 LOC101928371 LOC101928403 TEX37 RPIA MIR4436A |
dgv61n16 | Chr.2:87343737 - 90254660 on Build GRCh38 | Deletion | IGKV3D-15 IGKV2-24 LINC00152 IGKV1D-13 THNSL2 IGKV1D-8 IGKV3-20 LOC107985911 MIR4436A PLGLB2 ANKRD36BP2 EIF2AK3 TEX37 KRCC1 IGK RPIA LOC730268 FABP1 RGPD2 LOC101927050 IGKV1-5 IGKC LOC101928371 LOC101928403 MIR4435-1 MIR4780 IGKV2-29 SMYD1 FOXI3 IGKV2-26 |
dgv6947n54 | Chr.2:87815738 - 88850700 on Build GRCh38 | Gain | LOC101928371 LOC101928403 THNSL2 MIR4780 MIR4436A SMYD1 ANKRD36BP2 FOXI3 EIF2AK3 TEX37 KRCC1 RPIA FABP1 RGPD2 |
nsv1072005 | Chr.2:87360367 - 90256528 on Build GRCh38 | Deletion | IGKV3D-15 IGKV2-24 LINC00152 IGKV1D-13 THNSL2 IGKV1D-8 IGKV3-20 LOC107985911 MIR4436A PLGLB2 ANKRD36BP2 EIF2AK3 TEX37 KRCC1 IGK RPIA LOC730268 FABP1 RGPD2 LOC101927050 IGKV1-5 IGKC LOC101928371 LOC101928403 MIR4435-1 MIR4780 IGKV2-29 SMYD1 FOXI3 IGKV2-26 |
nsv1130752 | Chr.2:87360364 - 90256525 on Build GRCh38 | Deletion | IGKV3D-15 IGKV2-24 LINC00152 IGKV1D-13 THNSL2 IGKV1D-8 IGKV3-20 LOC107985911 MIR4436A PLGLB2 ANKRD36BP2 EIF2AK3 TEX37 KRCC1 IGK RPIA LOC730268 FABP1 RGPD2 LOC101927050 IGKV1-5 IGKC LOC101928371 LOC101928403 MIR4435-1 MIR4780 IGKV2-29 SMYD1 FOXI3 IGKV2-26 |
nsv582384 | Chr.2:87987149 - 88773214 on Build GRCh38 | Loss | LOC101928371 LOC101928403 THNSL2 MIR4780 SMYD1 ANKRD36BP2 FOXI3 EIF2AK3 TEX37 KRCC1 RPIA FABP1 RGPD2 |
nsv582386 | Chr.2:88035188 - 89208087 on Build GRCh38 | Loss | IGKV1-5 IGKC LOC101928371 IGKV2-24 LOC101928403 THNSL2 IGKV3-20 MIR4780 MIR4436A SMYD1 ANKRD36BP2 FOXI3 EIF2AK3 TEX37 KRCC1 IGK RPIA FABP1 IGKV2-26 |