Assay Details
Target Gene Details
Entrez Gene ID: | 57007 |
Gene Name: | atypical chemokine receptor 3 |
Gene Aliases: |
CMKOR1, CXC-R7, CXCR-7, CXCR7, GPR159, RDC-1, RDC1 |
Location: |
Chr.2:236537131-236582358 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ACKR3 | XM_005246098.3 | XP_005246155.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1005677 | Chr.2:236369512 - 236811991 on Build GRCh38 | Gain | ACKR3 IQCA1 LOC105373991 |
nsv997438 | Chr.2:236210757 - 237034862 on Build GRCh38 | Gain | ACKR3 ASB18 IQCA1 LOC105373991 |
nsv1000252 | Chr.2:235899999 - 238766417 on Build GRCh38 | Loss | ESPNL MIR6811 UBE2F-SCLY IQCA1 PRLH RAB17 LOC105373991 LOC105373944 KLHL30 TRAF3IP1 LRRFIP1 LOC93463 HES6 LINC01107 ASB1 GBX2 LOC643387 COL6A3 LOC151174 ACKR3 ERFE UBE2F SCLY ASB18 COPS8 RAMP1 MLPH PER2 LOC101927958 AGAP1 ILKAP LOC105373958 RBM44 |
nsv834576 | Chr.2:236466567 - 236654210 on Build GRCh38 | Gain | ACKR3 IQCA1 LOC105373991 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |