Assay Details
Target Gene Details
Entrez Gene ID: | 8443 |
Gene Name: | glyceronephosphate O-acyltransferase |
Gene Aliases: |
DAP-AT, DAPAT, DHAPAT, RCDP2 |
Location: |
Chr.1:231241173-231277973 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
GNPAT | NM_001316350.1 | NP_001303279.1 | ||
NM_014236.3 | NP_055051.1 | |||
XM_005273313.4 | XP_005273370.1 | |||
XM_011544303.2 | XP_011542605.1 | |||
XM_011544304.2 | XP_011542606.1 | |||
AB209882.1 | BAD93119.1 | |||
AF043937.1 | AAC24505.1 | |||
AJ002190.1 | CAA05242.1 | |||
AK222773.1 | BAD96493.1 | |||
AK223304.1 | BAD97024.1 | |||
AK297982.1 | ||||
AK309994.1 | ||||
AK311386.1 | ||||
AK314181.1 | ||||
BC000450.2 | AAH00450.1 | |||
DA552987.1 | ||||
DC366380.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3589106 | Chr.1:231110289 - 231278336 on Build GRCh38 | Gain | LOC149373 C1orf131 TRIM67 GNPAT |
esv3589093 | Chr.1:230482037 - 231277190 on Build GRCh38 | Gain | COG2 LOC101927604 MIR1182 FAM89A TTC13 C1orf131 LOC101927553 AGT TRIM67 ARV1 CAPN9 LOC149373 C1orf198 GNPAT |
nsv1000488 | Chr.1:230799658 - 231269735 on Build GRCh38 | Gain | LOC101927604 CAPN9 MIR1182 FAM89A TTC13 LOC149373 C1orf131 C1orf198 TRIM67 ARV1 GNPAT |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |