Assay Details
Target Gene Details
Entrez Gene ID: | 164045 |
Gene Name: | HFM1, ATP dependent DNA helicase homolog |
Gene Aliases: |
MER3, POF9, SEC63D1, Si-11, Si-11-6, helicase |
Location: |
Chr.1:91260766-91408008 on Build GRCh38 |
Assay Gene Location: | Within Intron 34 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
HFM1 | NM_001017975.4 | NP_001017975.4 | ||
XM_011540849.1 | XP_011539151.1 | |||
XM_011540850.2 | XP_011539152.1 | |||
XM_011540851.1 | XP_011539153.1 | |||
XM_011540852.2 | XP_011539154.1 | |||
XM_011540855.1 | XP_011539157.1 | |||
XM_011540857.1 | XP_011539159.1 | |||
XM_011540859.2 | XP_011539161.1 | |||
XM_017000490.1 | XP_016855979.1 | |||
XM_017000491.1 | XP_016855980.1 | |||
XM_017000492.1 | XP_016855981.1 | |||
XM_017000493.1 | XP_016855982.1 | |||
AB044787.1 | ||||
AB074274.1 | ||||
AB204867.1 | ||||
AK094079.1 | BAC04281.1 | |||
AK294754.1 | ||||
AK302582.1 | ||||
BC144172.1 | ||||
BX649037.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2714684 | Chr.1:91095800 - 93049130 on Build GRCh38 | Deletion | BRDT C1orf146 CDC7 EVI5 RPL5 FAM69A GLMN SNORD21 KIAA1107 GFI1 EPHX4 SNORA66 HSP90B3P SETSIP BTBD8 TGFBR3 HFM1 RPAP2 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |