Assay Details
Target Gene Details
Entrez Gene ID: | 2786 |
Gene Name: | G protein subunit gamma 4 |
Gene Aliases: |
- |
Location: |
Chr.1:235547685-235650754 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
GNG4 | NM_001098721.1 | NP_001092191.1 | ||
NM_001098722.1 | NP_001092192.1 | |||
NM_004485.3 | NP_004476.1 | |||
XM_006711761.2 | XP_006711824.1 | |||
XM_011544167.2 | XP_011542469.1 | |||
AF493872.1 | AAM12586.1 | |||
AK311781.1 | ||||
BC022485.1 | AAH22485.1 | |||
DA122711.1 | ||||
DA230013.1 | ||||
U31382.1 | AAC50204.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1013413 | Chr.1:235560712 - 235607405 on Build GRCh38 | Gain | GNG4 |
nsv1013571 | Chr.1:235210731 - 235587805 on Build GRCh38 | Gain | TBCE B3GALNT2 GGPS1 ARID4B GNG4 |
dgv30n21 | Chr.1:235198002 - 235605078 on Build GRCh38 | Gain | TBCE B3GALNT2 GGPS1 ARID4B GNG4 |
nsv1011288 | Chr.1:235376656 - 235666302 on Build GRCh38 | Gain | TBCE B3GALNT2 LYST GNG4 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |