Assay Details
Target Gene Details
Entrez Gene ID: | 5931 |
Gene Name: | RB binding protein 7, chromatin remodeling factor |
Gene Aliases: |
RbAp46 |
Location: |
Chr.X:16844650-16870411 on Build GRCh38 |
Assay Gene Location: | Within Intron 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
RBBP7 | NM_001198719.1 | NP_001185648.1 | ||
NM_002893.3 | NP_002884.1 | |||
AK091911.1 | ||||
AK127332.1 | ||||
AK291567.1 | ||||
BC114500.1 | ||||
BC114501.1 | ||||
BQ775345.1 | ||||
CR541701.1 | CAG46502.1 | |||
U35143.1 | AAC50231.1 | |||
X72841.1 | CAA51360.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic non-DGV Variation |