Assay Details
Target Gene Details
Entrez Gene ID: | 164633 |
Gene Name: | calcium binding protein 7 |
Gene Aliases: |
CALN2 |
Location: |
Chr.22:29720090-29731833 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CABP7 | NM_182527.2 | NP_872333.1 | ||
XM_006724161.3 | XP_006724224.1 | |||
BC051805.1 | AAH51805.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv471194 | Chr.22:29707487 - 29732391 on Build GRCh38 | Gain | ZMAT5 CABP7 |
nsv515720 | Chr.22:29685726 - 29767537 on Build GRCh38 | Gain+Loss | ZMAT5 NF2 UQCR10 CABP7 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |