Assay Details
Target Gene Details
Entrez Gene ID: | 10317 |
Gene Name: | beta-1,3-galactosyltransferase 5 |
Gene Aliases: |
B3GalT-V, B3GalTx, B3T5, GLCT5, beta-1,3-GalTase 5, beta-3-Gx-T5, beta3Gal-T5 |
Location: |
Chr.21:39612940-39662889 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv587485 | Chr.21:39628426 - 39656270 on Build GRCh38 | Loss | B3GALT5 |
nsv1061454 | Chr.21:39386047 - 39893479 on Build GRCh38 | Gain | MIR6508 IGSF5 B3GALT5-AS1 LCA5L WRB-SH3BGR WRB SH3BGR B3GALT5 PCP4 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |