Assay Details
Target Gene Details
Entrez Gene ID: | 8711 |
Gene Name: | tyrosine kinase non receptor 1 |
Gene Aliases: |
- |
Location: |
Chr.17:7380559-7389774 on Build GRCh38 |
Assay Gene Location: | Within Intron 11 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TNK1 | NM_001251902.1 | NP_001238831.1 | ||
NM_003985.4 | NP_003976.2 | |||
XM_011524045.2 | XP_011522347.1 | |||
XM_011524046.2 | XP_011522348.1 | |||
XM_011524047.2 | XP_011522349.1 | |||
XM_011524048.1 | XP_011522350.1 | |||
AK225403.1 | ||||
AK310684.1 | ||||
BC035782.1 | AAH35782.1 | |||
U43408.1 | AAC50427.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2422288 | Chr.17:6789933 - 7646709 on Build GRCh38 | Deletion | BCL6B ELP5 SOX15 TMEM256-PLSCR3 EIF5A NEURL4 CLDN7 SPEM1 CD68 LOC107983988 KCTD11 TEKT1 SNORA67 MPDU1 RNASEK YBX2 FBXO39 CLEC10A FXR2 TNK1 ACADVL SNORD10 TMEM102 TNFSF12 LOC100996842 TMEM95 EIF4A1 SAT2 ATP1B2 SLC35G6 SLC2A4 SLC16A11 TNFSF12-TNFSF13 ALOX12P2 GABARAP C17orf49 CTDNEP1 DLG4 MIR195 CHRNB1 TMEM256 GPS2 DVL2 SNORA48 SLC16A13 PLSCR3 ASGR2 RNASEK-C17orf49 MIR497HG FGF11 ACAP1 ALOX12-AS1 MIR497 TNFSF13 SENP3-EIF4A1 NLGN2 ALOX12 C17orf74 ZBTB4 PHF23 MIR324 POLR2A ASGR1 SENP3 SHBG |
nsv523672 | Chr.17:7379825 - 7549660 on Build GRCh38 | Loss | CHRNB1 NLGN2 TMEM256 TNK1 TMEM256-PLSCR3 C17orf74 ZBTB4 TMEM102 TNFSF12 SPEM1 PLSCR3 SLC35G6 TNFSF12-TNFSF13 LOC107983988 POLR2A FGF11 |
nsv952118 | Chr.17:7174582 - 7456981 on Build GRCh38 | Deletion | ELP5 CHRNB1 TMEM256 TMEM256-PLSCR3 EIF5A GPS2 NEURL4 DVL2 CLDN7 SPEM1 PLSCR3 LOC107983988 KCTD11 FGF11 ACAP1 YBX2 NLGN2 TNK1 C17orf74 ACADVL PHF23 TMEM102 MIR324 TMEM95 SLC2A4 GABARAP CTDNEP1 ASGR1 DLG4 |
nsv574322 | Chr.17:7010333 - 7689462 on Build GRCh38 | Loss | BCL6B ELP5 SOX15 TMEM256-PLSCR3 EIF5A NEURL4 CLDN7 SPEM1 CD68 LOC107983988 KCTD11 SNORA67 MPDU1 RNASEK YBX2 WRAP53 CLEC10A FXR2 TNK1 ACADVL SNORD10 TMEM102 TNFSF12 LOC100996842 TMEM95 EIF4A1 SAT2 ATP1B2 SLC35G6 SLC2A4 SLC16A11 TNFSF12-TNFSF13 GABARAP C17orf49 CTDNEP1 DLG4 MIR195 CHRNB1 TMEM256 GPS2 DVL2 SNORA48 SLC16A13 PLSCR3 ASGR2 RNASEK-C17orf49 MIR497HG FGF11 ACAP1 ALOX12-AS1 MIR497 TNFSF13 SENP3-EIF4A1 NLGN2 ALOX12 C17orf74 ZBTB4 PHF23 TP53 MIR324 POLR2A ASGR1 SENP3 SHBG |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |