Assay Details
Target Gene Details
Entrez Gene ID: | 81577 |
Gene Name: | glucose-fructose oxidoreductase domain containing 2 |
Gene Aliases: |
- |
Location: |
Chr.16:67674533-67719370 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
GFOD2 | NM_001243650.1 | NP_001230579.1 | ||
NM_030819.3 | NP_110446.3 | |||
NR_027398.1 | ||||
XM_017023738.1 | XP_016879227.1 | |||
AK074382.1 | BAB85066.1 | |||
AK225997.1 | ||||
AK307280.1 | ||||
BC000757.2 | AAH00757.1 | |||
BC107585.1 | AAI07586.1 | |||
BI826603.1 | ||||
DC368734.1 | ||||
DC381447.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv522852 | Chr.16:67153308 - 68071571 on Build GRCh38 | Gain | C16orf86 NRN1L GFOD2 RANBP10 EXOC3L1 LRRC29 CTRL TSNAXIP1 HSF4 TMEM208 ZDHHC1 PLEKHG4 SLC12A4 CTCF LOC107984813 AGRP LOC100131303 ELMO3 MIR328 FBXL8 FHOD1 TRADD THAP11 LOC105369155 EDC4 CENPT PSKH1 LCAT DPEP2 NUTF2 ATP6V0D1 E2F4 KIAA0895L ENKD1 LOC100505942 ACD TPPP3 PSMB10 NOL3 DDX28 CARMIL2 HSD11B2 PARD6A KCTD19 LRRC36 DPEP3 SLC9A5 DUS2 FAM65A |
nsv524363 | Chr.16:67136529 - 68071571 on Build GRCh38 | Loss | C16orf86 NRN1L GFOD2 RANBP10 EXOC3L1 LRRC29 CTRL TSNAXIP1 HSF4 TMEM208 ZDHHC1 B3GNT9 PLEKHG4 SLC12A4 CTCF LOC107984813 AGRP LOC100131303 ELMO3 MIR328 FBXL8 FHOD1 TRADD THAP11 LOC105369155 EDC4 CENPT PSKH1 LCAT DPEP2 NUTF2 ATP6V0D1 C16orf70 E2F4 KIAA0895L ENKD1 LOC100505942 ACD TPPP3 PSMB10 NOL3 DDX28 CARMIL2 HSD11B2 PARD6A KCTD19 LRRC36 DPEP3 SLC9A5 DUS2 FAM65A |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |