Assay Details
Target Gene Details
Entrez Gene ID: | 81577 |
Gene Name: | glucose-fructose oxidoreductase domain containing 2 |
Gene Aliases: |
- |
Location: |
Chr.16:67674533-67719370 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
GFOD2 | NM_001243650.1 | NP_001230579.1 | ||
NM_030819.3 | NP_110446.3 | |||
NR_027398.1 | ||||
AK074382.1 | BAB85066.1 | |||
AK225997.1 | ||||
AK307280.1 | ||||
BC000757.2 | AAH00757.1 | |||
BC107585.1 | AAI07586.1 | |||
BI826603.1 | ||||
DC368734.1 | ||||
DC381447.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv522852 | Chr.16:67153308 - 68071571 on Build GRCh38 | Gain | LOC100131303 ACD PSKH1 PLEKHG4 LOC107984813 HSF4 PSMB10 ATP6V0D1 DUS2 CTRL ZDHHC1 EDC4 C16orf86 LRRC36 NRN1L AGRP LCAT MIR328 TSNAXIP1 SLC9A5 ELMO3 TPPP3 NUTF2 LRRC29 HSD11B2 DDX28 CTCF FBXL8 LOC105369155 EXOC3L1 KIAA0895L TMEM208 LOC100505942 KCTD19 NOL3 FHOD1 PARD6A CENPT DPEP2 SLC12A4 FAM65A RANBP10 THAP11 DPEP3 ENKD1 GFOD2 TRADD E2F4 CARMIL2 |
nsv524363 | Chr.16:67136529 - 68071571 on Build GRCh38 | Loss | LOC100131303 ACD PSKH1 PLEKHG4 LOC107984813 HSF4 PSMB10 ATP6V0D1 DUS2 CTRL ZDHHC1 EDC4 C16orf86 LRRC36 NRN1L AGRP LCAT MIR328 TSNAXIP1 SLC9A5 ELMO3 TPPP3 NUTF2 LRRC29 HSD11B2 DDX28 CTCF FBXL8 LOC105369155 B3GNT9 EXOC3L1 KIAA0895L TMEM208 LOC100505942 KCTD19 NOL3 FHOD1 PARD6A CENPT DPEP2 SLC12A4 FAM65A C16orf70 RANBP10 THAP11 DPEP3 ENKD1 GFOD2 TRADD E2F4 CARMIL2 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |