Assay Details
Target Gene Details
Entrez Gene ID: | 81577 |
Gene Name: | glucose-fructose oxidoreductase domain containing 2 |
Gene Aliases: |
- |
Location: |
Chr.16:67674533-67719370 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
GFOD2 | NM_001243650.1 | NP_001230579.1 | ||
NM_030819.3 | NP_110446.3 | |||
NR_027398.1 | ||||
AK074382.1 | BAB85066.1 | |||
AK225997.1 | ||||
AK307280.1 | ||||
BC000757.2 | AAH00757.1 | |||
BC107585.1 | AAI07586.1 | |||
BI826603.1 | ||||
DC368734.1 | ||||
DC381447.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv522852 | Chr.16:67153308 - 68071571 on Build GRCh38 | Gain | DDX28 ELMO3 LRRC29 PARD6A EDC4 TRADD C16orf86 CTRL LOC107984813 THAP11 ATP6V0D1 LCAT ZDHHC1 PSKH1 FAM65A PLEKHG4 SLC9A5 CARMIL2 AGRP ENKD1 NOL3 TSNAXIP1 LRRC36 CENPT DPEP2 FBXL8 DUS2 HSF4 MIR328 FHOD1 E2F4 TPPP3 DPEP3 PSMB10 HSD11B2 LOC105369155 RANBP10 LOC100505942 KCTD19 GFOD2 NRN1L ACD NUTF2 KIAA0895L EXOC3L1 CTCF TMEM208 LOC100131303 SLC12A4 |
nsv524363 | Chr.16:67136529 - 68071571 on Build GRCh38 | Loss | DDX28 C16orf70 ELMO3 LRRC29 PARD6A EDC4 TRADD C16orf86 CTRL LOC107984813 THAP11 ATP6V0D1 LCAT ZDHHC1 PSKH1 FAM65A PLEKHG4 SLC9A5 B3GNT9 CARMIL2 AGRP ENKD1 NOL3 TSNAXIP1 LRRC36 CENPT DPEP2 FBXL8 DUS2 HSF4 MIR328 FHOD1 E2F4 TPPP3 DPEP3 PSMB10 HSD11B2 LOC105369155 RANBP10 LOC100505942 KCTD19 GFOD2 NRN1L ACD NUTF2 KIAA0895L EXOC3L1 CTCF TMEM208 LOC100131303 SLC12A4 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |