Assay Details
Target Gene Details
Entrez Gene ID: | 9333 |
Gene Name: | transglutaminase 5 |
Gene Aliases: |
PSS2, TGASE5, TGASEX, TGM6, TGMX, TGX |
Location: |
Chr.15:43232595-43266857 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TGM5 | NM_004245.3 | NP_004236.1 | ||
NM_201631.3 | NP_963925.2 | |||
AF035960.1 | AAC02978.1 | |||
AF035961.1 | AAC02979.1 | |||
AK298479.1 | ||||
BC119009.1 | ||||
BC122859.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic non-DGV Variation |