Assay Details
Target Gene Details
Entrez Gene ID: | 321 |
Gene Name: | amyloid beta precursor protein binding family A member 2 |
Gene Aliases: |
D15S1518E, HsT16821, LIN-10, MGC:14091, MINT2, X11-BETA, X11L |
Location: |
Chr.15:28885479-29118315 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
APBA2 | XM_011521489.2 | XP_011519791.1 | ||
XM_011521490.2 | XP_011519792.1 | |||
XM_011521493.2 | XP_011519795.1 | |||
XM_011521494.2 | XP_011519796.1 | |||
XM_017022101.1 | XP_016877590.1 | |||
XM_017022102.1 | XP_016877591.1 | |||
XM_017022105.1 | XP_016877594.1 | |||
XM_017022106.1 | XP_016877595.1 | |||
XM_017022107.1 | XP_016877596.1 | |||
XM_017022108.1 | XP_016877597.1 | |||
XM_017022109.1 | XP_016877598.1 | |||
XM_017022110.1 | XP_016877599.1 | |||
XM_017022113.1 | XP_016877602.1 | |||
XM_017022114.1 | XP_016877603.1 | |||
AK310413.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1040778 | Chr.15:28689598 - 30515727 on Build GRCh38 | Loss | GOLGA8J WHAMMP2 ULK4P3 LOC101060569 LOC100289656 LOC107984752 LOC100130111 TJP1 LOC105370746 GOLGA8T LOC105370751 NSMCE3 APBA2 GOLGA8M DKFZP434L187 GOLGA8R GOLGA6L7P CHRFAM7A FAM189A1 PDCD6IPP2 |
nsv527255 | Chr.15:28754022 - 30074044 on Build GRCh38 | Gain | WHAMMP2 GOLGA6L7P LOC100289656 LOC107984752 FAM189A1 PDCD6IPP2 LOC100130111 TJP1 NSMCE3 APBA2 |
esv2751526 | Chr.15:28724519 - 30687025 on Build GRCh38 | Loss | LOC100996413 GOLGA8J LOC101060569 LOC107984752 LOC100130111 LOC105370746 GOLGA8T LOC101927579 DKFZP434L187 ULK4P2 GOLGA6L7P CHRFAM7A FAM189A1 PDCD6IPP2 WHAMMP2 ULK4P3 LOC100289656 LOC100288637 TJP1 LOC105370751 NSMCE3 APBA2 GOLGA8R ARHGAP11B GOLGA8H |
nsv1039798 | Chr.15:28724126 - 30158154 on Build GRCh38 | Gain | GOLGA8J WHAMMP2 ULK4P3 LOC100289656 LOC107984752 LOC100130111 TJP1 GOLGA8T NSMCE3 APBA2 GOLGA6L7P FAM189A1 PDCD6IPP2 |
nsv568662 | Chr.15:27726493 - 29056123 on Build GRCh38 | Gain | WHAMMP2 LOC100289656 LOC107984752 GOLGA8G HERC2P9 OCA2 APBA2 LOC645202 GOLGA8M MIR4509-2 GOLGA8F LOC100132202 GOLGA6L7P HERC2P11 PDCD6IPP2 MIR4509-3 LOC101929047 HERC2 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |