Assay Details
Target Gene Details
Entrez Gene ID: | 8854 |
Gene Name: | aldehyde dehydrogenase 1 family member A2 |
Gene Aliases: |
RALDH(II), RALDH2, RALDH2-T |
Location: |
Chr.15:57953424-58065923 on Build GRCh38 |
Assay Gene Location: | Within Intron 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ALDH1A2 | NM_001206897.1 | NP_001193826.1 | ||
NM_003888.3 | NP_003879.2 | |||
NM_170696.2 | NP_733797.1 | |||
NM_170697.2 | NP_733798.1 | |||
AB015226.1 | BAA34785.1 | |||
AB015227.1 | BAA34786.1 | |||
AB015228.1 | BAA34787.1 | |||
AK128709.1 | ||||
AK294981.1 | ||||
AK303057.1 | ||||
BC030589.2 | AAH30589.1 | |||
BX443600.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv569593 | Chr.15:57909500 - 58016452 on Build GRCh38 | Loss | ALDH1A2 |
nsv833023 | Chr.15:57869641 - 58047976 on Build GRCh38 | Gain | ALDH1A2 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |