Assay Details
Target Gene Details
Entrez Gene ID: | 321 |
Gene Name: | amyloid beta precursor protein binding family A member 2 |
Gene Aliases: |
D15S1518E, HsT16821, LIN-10, MGC:14091, MINT2, X11-BETA, X11L |
Location: |
Chr.15:28885479-29118315 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1040778 | Chr.15:28689598 - 30515727 on Build GRCh38 | Loss | LOC105370746 GOLGA8M LOC100289656 FAM189A1 LOC105370751 PDCD6IPP2 CHRFAM7A LOC100130111 GOLGA8R NSMCE3 GOLGA6L7P WHAMMP2 APBA2 LOC107984752 ULK4P3 TJP1 DKFZP434L187 GOLGA8T GOLGA8J LOC101060569 |
nsv527255 | Chr.15:28754022 - 30074044 on Build GRCh38 | Gain | LOC100289656 GOLGA6L7P FAM189A1 PDCD6IPP2 WHAMMP2 APBA2 LOC107984752 TJP1 LOC100130111 NSMCE3 |
esv2751526 | Chr.15:28724519 - 30687025 on Build GRCh38 | Loss | LOC101927579 CHRFAM7A LOC100996413 LOC100130111 GOLGA8R NSMCE3 WHAMMP2 APBA2 DKFZP434L187 LOC105370746 LOC100289656 ARHGAP11B FAM189A1 LOC105370751 PDCD6IPP2 GOLGA8H LOC100288637 GOLGA6L7P LOC107984752 ULK4P3 TJP1 ULK4P2 GOLGA8T GOLGA8J LOC101060569 |
nsv1039798 | Chr.15:28724126 - 30158154 on Build GRCh38 | Gain | LOC100289656 FAM189A1 PDCD6IPP2 LOC100130111 NSMCE3 GOLGA6L7P WHAMMP2 APBA2 LOC107984752 ULK4P3 TJP1 GOLGA8T GOLGA8J |
nsv516384 | Chr.15:28920744 - 28996217 on Build GRCh38 | Loss | APBA2 |
nsv1038806 | Chr.15:28933682 - 28965543 on Build GRCh38 | Gain | APBA2 |
nsv568662 | Chr.15:27726493 - 29056123 on Build GRCh38 | Gain | GOLGA8M LOC100289656 OCA2 PDCD6IPP2 LOC100132202 MIR4509-2 GOLGA6L7P HERC2 GOLGA8F WHAMMP2 APBA2 LOC107984752 LOC645202 HERC2P11 HERC2P9 LOC101929047 GOLGA8G MIR4509-3 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |