Assay Details
Target Gene Details
Entrez Gene ID: | 10060 |
Gene Name: | ATP binding cassette subfamily C member 9 |
Gene Aliases: |
ABC37, ATFB12, CANTU, CMD1O, SUR2 |
Location: |
Chr.12:21797389-21941869 on Build GRCh38 |
Assay Gene Location: | Within Intron 38 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ABCC9 | NM_005691.3 | NP_005682.2 | ||
NM_020297.3 | NP_064693.2 | |||
XM_005253284.3 | XP_005253341.1 | |||
XM_005253286.3 | XP_005253343.1 | |||
XM_005253287.4 | XP_005253344.1 | |||
XM_005253288.3 | XP_005253345.1 | |||
XM_005253289.3 | XP_005253346.1 | |||
XM_005253290.3 | XP_005253347.1 | |||
XM_006719025.3 | XP_006719088.1 | |||
XM_011520545.2 | XP_011518847.1 | |||
AK092535.1 | ||||
AK094171.1 | BAC04300.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv557741 | Chr.12:21425625 - 22016920 on Build GRCh38 | Gain | ABCC9 LDHB PYROXD1 SPX GYS2 GOLT1B KCNJ8 RECQL |
nsv557744 | Chr.12:21535966 - 21813767 on Build GRCh38 | Loss | ABCC9 LDHB GYS2 KCNJ8 |
nsv470275 | Chr.12:21785610 - 21973380 on Build GRCh38 | Gain | ABCC9 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |