Assay Details
Target Gene Details
Entrez Gene ID: | 100526737 |
Gene Name: | RBM14-RBM4 readthrough |
Gene Aliases: |
COAZ |
Location: |
Chr.11:66616582-66646473 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
RBM14-RBM4 | NM_001198845.1 | NP_001185774.1 | ||
NM_001198846.1 | NP_001185775.1 | |||
AW958943.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv522277 | Chr.11:66466552 - 66726354 on Build GRCh38 | Loss | SPTBN2 CCDC87 PELI3 BBS1 DPP3 CCS RBM4 ACTN3 RBM14-RBM4 RBM4B RBM14 ZDHHC24 LOC101928069 CTSF |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |