Assay Details
Target Gene Details
Entrez Gene ID: | 9645 |
Gene Name: | microtubule associated monooxygenase, calponin and LIM domain containing 2 |
Gene Aliases: |
MICAL-2, MICAL2PV1, MICAL2PV2 |
Location: |
Chr.11:12110576-12281017 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
MICAL2 | NM_001282663.1 | NP_001269592.1 | ||
NM_001282668.1 | NP_001269597.1 | |||
NM_014632.3 | NP_055447.1 | |||
AB018293.1 | ||||
AK302580.1 | ||||
AK302893.1 | ||||
BC044577.1 | AAH44577.1 | |||
DA246062.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv483000 | Chr.11:10678454 - 12678453 on Build GRCh38 | Loss | LOC105376554 LOC101928053 PARVA SNORD97 MICALCL MICAL2 MIR6124 EIF4G2 GALNT18 DKK3 MRVI1 MIR8070 LOC105376545 ZBED5-AS1 ZBED5 USP47 TEAD1 MIR4299 CSNK2A3 CTR9 |
nsv832068 | Chr.11:12098790 - 12284080 on Build GRCh38 | Gain | MICAL2 MIR6124 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |