Assay Details
Target Gene Details
Entrez Gene ID: | 51063 |
Gene Name: | calcium homeostasis modulator 2 |
Gene Aliases: |
FAM26B |
Location: |
Chr.10:103446785-103452419 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CALHM2 | NM_015916.4 | NP_057000.2 | ||
NR_024552.1 | ||||
NR_046344.1 | ||||
XM_006717880.2 | XP_006717943.1 | |||
XM_006717883.2 | XP_006717946.1 | |||
XM_006717884.3 | XP_006717947.1 | |||
XM_011539848.2 | XP_011538150.1 | |||
XM_017016306.1 | XP_016871795.1 | |||
XM_017016307.1 | XP_016871796.1 | |||
XM_017016308.1 | XP_016871797.1 | |||
XM_017016309.1 | XP_016871798.1 | |||
AF131810.1 | AAD20050.1 | |||
AK022195.1 | BAB13983.1 | |||
AK125276.1 | ||||
BC000039.2 | AAH00039.1 | |||
DB109582.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv524966 | Chr.10:103441820 - 103717405 on Build GRCh38 | Loss | CALHM3 PDCD11 NEURL1 CALHM2 CALHM1 NEURL1-AS1 SH3PXD2A |
nsv518987 | Chr.10:103434329 - 103484808 on Build GRCh38 | Loss | CALHM3 PDCD11 CALHM2 CALHM1 NEURL1-AS1 |
dgv965n100 | Chr.10:103385173 - 103457152 on Build GRCh38 | Gain | TAF5 PDCD11 CALHM2 MIR1307 USMG5 CALHM1 |
dgv280e212 | Chr.10:103390805 - 103464349 on Build GRCh38 | Gain | PDCD11 CALHM2 MIR1307 USMG5 CALHM1 |
esv3577694 | Chr.10:103391276 - 103553473 on Build GRCh38 | Gain | CALHM3 PDCD11 NEURL1 CALHM2 MIR1307 USMG5 CALHM1 NEURL1-AS1 |
esv3624414 | Chr.10:103412843 - 103459270 on Build GRCh38 | Gain | PDCD11 CALHM2 CALHM1 |
nsv552060 | Chr.10:103447748 - 103470575 on Build GRCh38 | Loss | CALHM2 CALHM1 |
esv3624413 | Chr.10:103392636 - 103459758 on Build GRCh38 | Gain | PDCD11 CALHM2 MIR1307 USMG5 CALHM1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |