Assay Details
Target Gene Details
Entrez Gene ID: | 23560 |
Gene Name: | GTP binding protein 4 |
Gene Aliases: |
CRFG, NGB, NOG1 |
Location: |
Chr.10:988409-1017768 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
GTPBP4 | NM_012341.2 | NP_036473.2 | ||
AF120334.1 | AAD09830.1 | |||
AF325353.1 | AAK13444.1 | |||
AK001548.1 | ||||
AK001552.1 | BAA91752.1 | |||
AK097093.1 | ||||
AK222861.1 | BAD96581.1 | |||
AK295231.1 | ||||
AK301721.1 | ||||
AK302219.1 | ||||
BC033784.1 | AAH33784.1 | |||
BC038975.2 | AAH38975.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2660097 | Chr.10:526001 - 1246069 on Build GRCh38 | Deletion | GTPBP4 DIP2C LOC105376341 PRR26 WDR37 LOC101927762 LOC101930421 LARP4B IDI2 IDI1 LOC105376339 IDI2-AS1 MIR5699 LINC00200 ADARB2 |
esv3891735 | Chr.10:978274 - 1064524 on Build GRCh38 | Gain | GTPBP4 IDI2-AS1 WDR37 IDI2 IDI1 |
nsv549765 | Chr.10:688289 - 1083587 on Build GRCh38 | Gain | GTPBP4 DIP2C LOC105376341 IDI2-AS1 WDR37 LOC101927762 LARP4B IDI2 IDI1 |
nsv1161921 | Chr.10:823011 - 1033401 on Build GRCh38 | Duplication | GTPBP4 LOC105376341 IDI2-AS1 LOC101927762 LARP4B IDI2 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |