Assay Details
Target Gene Details
Entrez Gene ID: | 23522 |
Gene Name: | lysine acetyltransferase 6B |
Gene Aliases: |
GTPTS, MORF, MOZ2, MYST4, ZC2HC6B, qkf, querkopf |
Location: |
Chr.10:74824927-75032623 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
KAT6B | NM_001256468.1 | NP_001243397.1 | ||
NM_001256469.1 | NP_001243398.1 | |||
NM_012330.3 | NP_036462.2 | |||
XM_005269664.2 | XP_005269721.1 | |||
XM_017016000.1 | XP_016871489.1 | |||
XM_017016001.1 | XP_016871490.1 | |||
XM_017016002.1 | XP_016871491.1 | |||
XM_017016003.1 | XP_016871492.1 | |||
XM_017016004.1 | XP_016871493.1 | |||
XM_017016005.1 | XP_016871494.1 | |||
XM_017016006.1 | XP_016871495.1 | |||
XM_017016007.1 | XP_016871496.1 | |||
XM_017016008.1 | XP_016871497.1 | |||
XM_017016009.1 | XP_016871498.1 | |||
AF217500.1 | AAL56647.1 | |||
AK074816.1 | ||||
BC014143.2 | AAH14143.1 | |||
BC048199.1 | AAH48199.1 | |||
BC150618.1 | ||||
DA488537.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv551535 | Chr.10:74745813 - 74842047 on Build GRCh38 | Loss | KAT6B |
esv2674729 | Chr.10:74824387 - 74831334 on Build GRCh38 | Deletion | KAT6B |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |