Assay Details
Target Gene Details
Entrez Gene ID: | 9108 |
Gene Name: | myotubularin related protein 7 |
Gene Aliases: |
- |
Location: |
Chr.8:17296794-17413531 on Build GRCh38 |
Assay Gene Location: | Within Intron 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
MTMR7 | NM_004686.4 | NP_004677.3 | ||
XM_006716414.3 | XP_006716477.2 | |||
XM_011544692.2 | XP_011542994.1 | |||
XM_011544693.2 | XP_011542995.1 | |||
XM_017013956.1 | XP_016869445.1 | |||
AF073482.1 | AAC77820.1 | |||
AK294468.1 | ||||
AK316022.1 | ||||
AK316283.1 | ||||
BC130577.1 | ||||
BC130579.1 | ||||
CR749240.1 | CAH18096.1 |
Target Gene Details
Entrez Gene ID: | 137492 |
Gene Name: | VPS37A, ESCRT-I subunit |
Gene Aliases: |
HCRP1, PQBP2, SPG53 |
Location: |
Chr.8:17246892-17333532 on Build GRCh38 |
Assay Gene Location: | Within Intron 14 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
VPS37A | XM_017013022.1 | XP_016868511.1 | ||
XM_017013025.1 | XP_016868514.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv831245 | Chr.8:17255574 - 17450899 on Build GRCh38 | Gain | VPS37A MTMR7 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |