Assay Details
Target Gene Details
Entrez Gene ID: | 54904 |
Gene Name: | Wolf-Hirschhorn syndrome candidate 1-like 1 |
Gene Aliases: |
NSD3, WHISTLE, pp14328 |
Location: |
Chr.8:38275042-38382272 on Build GRCh38 |
Assay Gene Location: | Within Intron 15 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
WHSC1L1 | NM_023034.1 | NP_075447.1 | ||
AF332469.1 | AAK00355.1 | |||
AJ295990.1 | CAC28350.1 | |||
AJ295991.1 | CAC28351.1 | |||
BC101717.1 | AAI01718.1 | |||
BC113469.1 | ||||
BC115006.1 | AAI15007.1 | |||
BC143510.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
dgv7164n100 | Chr.8:38194164 - 38310837 on Build GRCh38 | Gain | BAG4 WHSC1L1 PLPP5 DDHD2 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |