Assay Details
Target Gene Details
Entrez Gene ID: | 114926 |
Gene Name: | small integral membrane protein 19 |
Gene Aliases: |
C8orf40 |
Location: |
Chr.8:42541155-42552997 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SMIM19 | NM_001135674.1 | NP_001129146.1 | ||
NM_001135675.1 | NP_001129147.1 | |||
NM_001135676.1 | NP_001129148.1 | |||
NM_138436.3 | NP_612445.2 | |||
XM_005273398.4 | XP_005273455.1 | |||
AK311932.1 | ||||
BC013035.2 | AAH13035.1 | |||
BI335341.1 | ||||
BI463786.1 | ||||
BI549696.1 | ||||
DA702519.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1017899 | Chr.8:42510362 - 42586679 on Build GRCh38 | Gain | SMIM19 SLC20A2 |