Assay Details
Target Gene Details
Entrez Gene ID: | 92421 |
Gene Name: | charged multivesicular body protein 4C |
Gene Aliases: |
SNF7-3, Shax3, VPS32C |
Location: |
Chr.8:81732453-81759515 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CHMP4C | NM_152284.3 | NP_689497.1 | ||
AB120734.1 | BAC87888.1 | |||
AK000049.1 | ||||
AK314873.1 | ||||
AY329086.1 | AAQ91195.1 | |||
BC014321.1 | AAH14321.1 | |||
BX492886.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv611607 | Chr.8:81405621 - 81768032 on Build GRCh38 | Gain | SLC10A5 FABP4 FABP12 FABP9 IMPA1 PMP2 CHMP4C ZFAND1 |
nsv831369 | Chr.8:81605833 - 81823586 on Build GRCh38 | Gain+Loss | SLC10A5 IMPA1 CHMP4C SNX16 ZFAND1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |