Assay Details
Target Gene Details
Entrez Gene ID: | 114 |
Gene Name: | adenylate cyclase 8 |
Gene Aliases: |
AC8, ADCY3, HBAC1 |
Location: |
Chr.8:130780300-131041604 on Build GRCh38 |
Assay Gene Location: | Within Intron 18 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ADCY8 | NM_001115.2 | NP_001106.1 | ||
XM_005250769.3 | XP_005250826.1 | |||
XM_006716501.3 | XP_006716564.1 | |||
XM_017013006.1 | XP_016868495.1 | |||
DQ104739.1 | ||||
GQ472223.1 | ||||
M83533.1 | AAA35523.2 | |||
Z35309.1 | CAA84552.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1016494 | Chr.8:130133708 - 131222639 on Build GRCh38 | Gain |
![]() ![]() ![]() |
nsv612294 | Chr.8:130632833 - 130911131 on Build GRCh38 | Gain |
![]() |
nsv831462 | Chr.8:130691873 - 130883547 on Build GRCh38 | Gain |
![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)