Assay Details
Target Gene Details
Entrez Gene ID: | 29960 |
Gene Name: | mitochondrial rRNA methyltransferase 2 |
Gene Aliases: |
FJH1, FTSJ2, HEL97, RRMJ2 |
Location: |
Chr.7:2234291-2242198 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
MRM2 | NM_013393.1 | NP_037525.1 | ||
AF093415.1 | AAF22488.1 | |||
AK302060.1 | ||||
BC114514.1 | AAI14515.1 | |||
BC114564.1 | AAI14565.1 | |||
CR749618.1 | CAH18412.1 | |||
EU794658.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1021310 | Chr.7:1936627 - 2280155 on Build GRCh38 | Loss | MIR6836 SNX8 MRM2 MAD1L1 NUDT1 |
nsv1018563 | Chr.7:2233301 - 2325297 on Build GRCh38 | Gain | MIR6836 SNX8 MRM2 NUDT1 |
nsv1021173 | Chr.7:2206456 - 2334992 on Build GRCh38 | Loss | MIR6836 SNX8 MRM2 MAD1L1 NUDT1 |
nsv830886 | Chr.7:2145358 - 2337903 on Build GRCh38 | Loss | MIR6836 SNX8 MRM2 MAD1L1 NUDT1 |
esv2761136 | Chr.7:2230260 - 2325309 on Build GRCh38 | Gain | MIR6836 SNX8 MRM2 MAD1L1 NUDT1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |