Assay Details
Target Gene Details
Entrez Gene ID: | 5445 |
Gene Name: | paraoxonase 2 |
Gene Aliases: |
- |
Location: |
Chr.7:95404862-95435072 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PON2 | NM_000305.2 | NP_000296.2 | ||
NM_001018161.1 | NP_001018171.1 | |||
XM_005250453.1 | XP_005250510.1 | |||
XM_017012357.1 | XP_016867846.1 | |||
XM_017012358.1 | XP_016867847.1 | |||
AB102891.1 | BAD89420.1 | |||
AF001601.1 | AAC27944.1 | |||
AF001602.1 | AAC27945.1 | |||
AK291103.1 | ||||
AK296029.1 | ||||
AK296636.1 | ||||
AK301313.1 | ||||
AK308065.1 | ||||
AK315209.1 | ||||
BC009728.2 | AAH09728.1 | |||
BC040010.1 | AAH40010.1 | |||
BC046160.1 | AAH46160.1 | |||
L48513.1 | AAC41995.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1023193 | Chr.7:93602376 - 95828660 on Build GRCh38 | Gain | PEG10 PON1 DYNC1I1 LOC105375401 PON2 PDK4 PPP1R9A GNGT1 LOC105375405 TFPI2 BET1 CASD1 GNG11 ASB4 PON3 COL1A2 MIR4652 SGCE |
esv3614194 | Chr.7:95410059 - 95421625 on Build GRCh38 | Gain | PON2 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |