Assay Details
Target Gene Details
Entrez Gene ID: | 9263 |
Gene Name: | serine/threonine kinase 17a |
Gene Aliases: |
DRAK1 |
Location: |
Chr.7:43583093-43627379 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
STK17A | NM_004760.2 | NP_004751.2 | ||
XM_017012792.1 | XP_016868281.1 | |||
AB011420.1 | BAA34126.1 | |||
BC023508.2 | AAH23508.2 | |||
BC047696.1 | AAH47696.1 | |||
BI461892.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3612997 | Chr.7:43585283 - 43592494 on Build GRCh38 | Loss | STK17A |
nsv606736 | Chr.7:43587460 - 43591529 on Build GRCh38 | Loss | STK17A |
nsv606717 | Chr.7:43564508 - 43592243 on Build GRCh38 | Loss | HECW1 STK17A |
esv3612993 | Chr.7:43438543 - 43609526 on Build GRCh38 | Gain | HECW1 LOC100506895 STK17A COA1 |
dgv11295n54 | Chr.7:43585908 - 43595683 on Build GRCh38 | Loss | STK17A |
esv2665229 | Chr.7:43585279 - 43592478 on Build GRCh38 | Deletion | STK17A |
esv3612996 | Chr.7:43537785 - 43656356 on Build GRCh38 | Gain | HECW1 STK17A COA1 |
esv22147 | Chr.7:43585305 - 43592406 on Build GRCh38 | Loss | STK17A |
dgv11292n54 | Chr.7:43583207 - 43592243 on Build GRCh38 | Loss | STK17A |
nsv606744 | Chr.7:43588950 - 43595683 on Build GRCh38 | Loss | STK17A |
dgv11296n54 | Chr.7:43588145 - 43592243 on Build GRCh38 | Loss | STK17A |
nsv518728 | Chr.7:43555801 - 43614160 on Build GRCh38 | Loss | HECW1 STK17A COA1 |
dgv1904e212 | Chr.7:43588031 - 43596574 on Build GRCh38 | Loss | STK17A |
esv3541556 | Chr.7:43584902 - 43592901 on Build GRCh38 | Deletion | STK17A |
dgv11293n54 | Chr.7:43583370 - 43604236 on Build GRCh38 | Loss | STK17A |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |